Order Entry
Poland
ContactUsLinkComponent
2-(5-Bromo-2-ethoxy-3-pyridyl)-6-methyl-1,3,6,2-dioxazaborocane-4,8-dione
2-(5-Bromo-2-ethoxy-3-pyridyl)-6-methyl-1,3,6,2-dioxazaborocane-4,8-dione
Catalog # APOSOR471521-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(5-Bromo-2-ethoxy-3-pyridyl)-6-methyl-1,3,6,2-dioxazaborocane-4,8-dione
Catalog # APOSOR471521-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :356.965 g/mol
Storage Temperature:Ambient
Formula:C₁₂H₁₄BBrN₂O₅
MDL Number:MFCD29049231