Order Entry
Poland
Orders LinkContactUsLinkComponent
1-(3,5-Dichlorophenyl)-3-(ethoxycarbonyl)urea

1-(3,5-Dichlorophenyl)-3-(ethoxycarbonyl)urea

Catalog #
1-(3,5-Dichlorophenyl)-3-(ethoxycarbonyl)urea
Catalog #
Supplier:  
CAS Number:  199741-78-1
1-(3,5-Dichlorophenyl)-3-(ethoxycarbonyl)urea
CAS Number:  199741-78-1
Product Details & Documents
  • CAS Number:199741-78-1
  • Formula:C₁₀H₁₀Cl₂N₂O₃
  • MDL Number:MFCD00135183
  • Molecular Weight:277.104 g/mol
  • Storage Temperature:Ambient