Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-(3-Piperidyl)propyl Acetate Acetate
3-(3-Piperidyl)propyl Acetate Acetate
3-(3-Piperidyl)propyl Acetate Acetate
3-(3-Piperidyl)propyl Acetate Acetate
APOSOR470598-5G
:
Apollo Scientific
:
3-(3-Piperidyl)propyl Acetate Acetate
APOSOR470598-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
245.315 g/mol
:
Ambient
:
1427475-20-4
:
C₁₀H₁₉NO₂·CH₃CO₂H
:
MFCD23381116