Order Entry
Poland
ContactUsLinkComponent
6-(3,5-Dichlorophenyl)-3-pyridinecarbaldehyde
6-(3,5-Dichlorophenyl)-3-pyridinecarbaldehyde
Catalog # APOSOR470364-1G
Supplier:  Apollo Scientific
CAS Number:  
6-(3,5-Dichlorophenyl)-3-pyridinecarbaldehyde
Catalog # APOSOR470364-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :252.096 g/mol
Storage Temperature:Ambient
CAS Number:898796-01-5
Formula:C₁₂H₇Cl₂NO
MDL Number:MFCD04115466