Order Entry
Poland
ContactUsLinkComponent
 
3-(2-Bromophenyl)-1,4-diazaspiro[4.4]nonan-2-one
Catalog # APOSOR470233-5G
Supplier:  Apollo Scientific
CAS Number:  
3-(2-Bromophenyl)-1,4-diazaspiro[4.4]nonan-2-one
Catalog # APOSOR470233-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :295.175 g/mol
Storage Temperature:Ambient
CAS Number:1272755-96-0
Formula:C₁₃H₁₅BrN₂O
MDL Number:MFCD18651741