Order Entry
Poland
ContactUsLinkComponent
4-Hydroxy-3-methoxybenzeneboronic acid 95+%
Irritant
4-Hydroxy-3-methoxybenzeneboronic acid 95+%
Catalog # APOSOR46236-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-Hydroxy-3-methoxybenzeneboronic acid 95+%
Catalog # APOSOR46236-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :167.955 g/mol
 :GHS07
Storage Temperature:Freezer
CAS Number:182344-21-4
Formula:C₇H₉BO₄
MDL Number:MFCD18397223