Order Entry
Poland
ContactUsLinkComponent
N-Boc-2-Azidoethylamine 97%
N-Boc-2-Azidoethylamine 97%
Catalog # APOSOR452182-1G
Supplier:  Apollo Scientific
CAS Number:  
N-Boc-2-Azidoethylamine 97%
Catalog # APOSOR452182-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :186.212 g/mol
Storage Temperature:Freezer
CAS Number:117861-38-8
Formula:C₇H₁₄N₄O₂
MDL Number:MFCD18207539