Order Entry
Poland
ContactUsLinkComponent
2,3,4,5-Tetrahydropyrazolo[1,5-b][1,2]oxazepine-7-carboxylic acid
2,3,4,5-Tetrahydropyrazolo[1,5-b][1,2]oxazepine-7-carboxylic acid
Catalog # APOSOR451247-250MG
Supplier:  Apollo Scientific
CAS Number:  
2,3,4,5-Tetrahydropyrazolo[1,5-b][1,2]oxazepine-7-carboxylic acid
Catalog # APOSOR451247-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :182.177 g/mol
Storage Temperature:Ambient
CAS Number:1934470-69-5
Formula:C₈H₁₀N₂O₃
MDL Number:MFCD28975211