Order Entry
Poland
ContactUsLinkComponent
(1H-Pyrrolo[2,3-c]pyridin-2-yl)methanol
Irritant
(1H-Pyrrolo[2,3-c]pyridin-2-yl)methanol
Catalog # APOSOR43671-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
(1H-Pyrrolo[2,3-c]pyridin-2-yl)methanol
Catalog # APOSOR43671-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :148.162 g/mol
Melting Pt:185 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:882881-15-4
Formula:C₈H₈N₂O
MDL Number:MFCD11506123