
3-Chloroimidazo[1,2-a]pyridine-2-carboxylic acid monohydrochloride monohydrate
Catalog # APOSOR40684-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:251.067 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:2022965-72-4
Formula:C₈H₅ClN₂O₂·ClH·H₂O
MDL Number:MFCD27995551