Order Entry
Poland
ContactUsLinkComponent
2-(Chloromethyl)-1-methyl-1H-imidazole hydrochloride 95%
Corrosive
2-(Chloromethyl)-1-methyl-1H-imidazole hydrochloride 95%
Catalog # APOSOR40445-250MG
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
2-(Chloromethyl)-1-methyl-1H-imidazole hydrochloride 95%
Catalog # APOSOR40445-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :167.036 g/mol
Melting Pt:175…178 °C
Storage Temperature:Ambient
UN:3261
ADR:8,III
 :GHS05
CAS Number:78667-04-6
Formula:C₅H₇ClN₂·ClH
MDL Number:MFCD00234118