
Methyl 2-(bromomethyl)-4-nitrobenzoate 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:133446-99-8
:
Formula:C₉H₈BrNO₄
MDL Number:MFCD11505948
:274.068 g/mol
Storage Temperature:Ambient
UN:3261
ADR:8,III
:GHS05
Frequently Bought Together