Order Entry
Poland
ContactUsLinkComponent
(5S)-tert-Butyl 3-bromo-5-methylpiperidine-1-carboxylate ≥98%
(5S)-tert-Butyl 3-bromo-5-methylpiperidine-1-carboxylate ≥98%
Catalog # APOSOR361728-100MG
Supplier:  Apollo Scientific
CAS Number:  
(5S)-tert-Butyl 3-bromo-5-methylpiperidine-1-carboxylate ≥98%
Catalog # APOSOR361728-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :278.186 g/mol
Storage Temperature:Ambient
CAS Number:2096990-26-8
Formula:C₁₁H₂₀BrNO₂
MDL Number:MFCD30728491