5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2-(2-((trimethylsilyl)oxy)propan-2-yl)pyridine
Catalog # APOSOR350390-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:335.321 g/mol
Storage Temperature:Ambient
CAS Number:1228014-10-5
Formula:C₁₇H₃₀BNO₃Si
MDL Number:MFCD12198135