Order Entry
Poland
ContactUsLinkComponent
4-Oxo-4,5,6,7-tetrahydro-pyrazolo[1,5-a]pyridine-2-carboxylic acid
4-Oxo-4,5,6,7-tetrahydro-pyrazolo[1,5-a]pyridine-2-carboxylic acid
Catalog # APOSOR346364-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Oxo-4,5,6,7-tetrahydro-pyrazolo[1,5-a]pyridine-2-carboxylic acid
Catalog # APOSOR346364-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :180.161 g/mol
Storage Temperature:Ambient
CAS Number:459157-20-1
Formula:C₈H₈N₂O₃
MDL Number:MFCD16658488