Order Entry
Poland
ContactUsLinkComponent
5,5-Dimethyl-2-phenyl-1,2,4-triazolidine-3-thione, tech
5,5-Dimethyl-2-phenyl-1,2,4-triazolidine-3-thione, tech
Catalog # APOSOR33424-1G
Supplier:  Apollo Scientific
CAS Number:  
5,5-Dimethyl-2-phenyl-1,2,4-triazolidine-3-thione, tech
Catalog # APOSOR33424-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :207.295 g/mol
Storage Temperature:Ambient
CAS Number:39263-68-8
Formula:C₁₀H₁₃N₃S
MDL Number:MFCD00684882