Order Entry
Poland
ContactUsLinkComponent
1-{6-[(2-Hydroxyethyl)(methyl)amino]naphthalen-2-yl}ethan-1-one 97%
1-{6-[(2-Hydroxyethyl)(methyl)amino]naphthalen-2-yl}ethan-1-one 97%
Catalog # APOSOR33301-1G
Supplier:  Apollo Scientific
CAS Number:  
1-{6-[(2-Hydroxyethyl)(methyl)amino]naphthalen-2-yl}ethan-1-one 97%
Catalog # APOSOR33301-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :243.301 g/mol
Storage Temperature:Ambient
CAS Number:259739-00-9
Formula:C₁₅H₁₇NO₂
MDL Number:MFCD12756735