Order Entry
Poland
ContactUsLinkComponent
(1E)-4-(4-Chlorophenoxy)-1-(dimethylamino)pent-1-en-3-one, tech
(1E)-4-(4-Chlorophenoxy)-1-(dimethylamino)pent-1-en-3-one, tech
Catalog # APOSOR33210-1G
Supplier:  Apollo Scientific
CAS Number:  
(1E)-4-(4-Chlorophenoxy)-1-(dimethylamino)pent-1-en-3-one, tech
Catalog # APOSOR33210-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:

Specifications

About this item

 :
 :253.725 g/mol
Storage Temperature:Ambient
CAS Number:1164506-63-1
Formula:C₁₃H₁₆ClNO₂
MDL Number:MFCD00139114