Methyl 2-(1-methyl-4-oxo-1,4-dihydroquinolin-3-yl)benzoate, tech
Catalog # APOSOR33048-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:293.317 g/mol
Storage Temperature:Ambient
CAS Number:294184-16-0
Formula:C₁₈H₁₅NO₃
MDL Number:MFCD05256228