Order Entry
Poland
ContactUsLinkComponent
2-(1-Methyl-4-oxo-1,4-dihydroquinolin-3-yl)benzoic acid, tech
2-(1-Methyl-4-oxo-1,4-dihydroquinolin-3-yl)benzoic acid, tech
Catalog # APOSOR32941-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(1-Methyl-4-oxo-1,4-dihydroquinolin-3-yl)benzoic acid, tech
Catalog # APOSOR32941-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :279.29 g/mol
Storage Temperature:Ambient
CAS Number:105576-79-2
Formula:C₁₇H₁₃NO₃
MDL Number:MFCD00207535