Order Entry
Poland
ContactUsLinkComponent
9-Amino-3,3-dimethyl-1,2,3,4-tetrahydroacridin-1-one, tech
9-Amino-3,3-dimethyl-1,2,3,4-tetrahydroacridin-1-one, tech
Catalog # APOSOR32875-1G
Supplier:  Apollo Scientific
CAS Number:  
9-Amino-3,3-dimethyl-1,2,3,4-tetrahydroacridin-1-one, tech
Catalog # APOSOR32875-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :240.3 g/mol
Storage Temperature:Ambient
CAS Number:130186-64-0
Formula:C₁₅H₁₆N₂O
MDL Number:MFCD00445032