Order Entry
Poland
ContactUsLinkComponent
(2Z)-3-[4-(Dimethylamino)phenyl]-2-[(1E)-(methoxyimino)methyl]prop-2-enenitrile, tech
(2Z)-3-[4-(Dimethylamino)phenyl]-2-[(1E)-(methoxyimino)methyl]prop-2-enenitrile, tech
Catalog # APOSOR32754-1G
Supplier:  Apollo Scientific
CAS Number:  
(2Z)-3-[4-(Dimethylamino)phenyl]-2-[(1E)-(methoxyimino)methyl]prop-2-enenitrile, tech
Catalog # APOSOR32754-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :229.278 g/mol
Storage Temperature:Ambient
CAS Number:339278-27-2
Formula:C₁₃H₁₅N₃O
MDL Number:MFCD00663698