2-Oxo-5-phenyl-1,2-dihydropyridine-3-carbonitrile, tech
Catalog # APOSOR32746-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:196.205 g/mol
Storage Temperature:Ambient
CAS Number:35982-93-5
Formula:C₁₂H₈N₂O
MDL Number:MFCD00231563