(2E)-1-(3-Chlorophenyl)-3-(dimethylamino)prop-2-en-1-one 95%
Catalog # APOSOR32732-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:209.672 g/mol
Storage Temperature:Ambient
CAS Number:876376-75-9
Formula:C₁₁H₁₂ClNO
MDL Number:MFCD00172143