Order Entry
Poland
ContactUsLinkComponent
(2Z)-2-[(Pyridin-3-yl)methylidene]-1-azabicyclo[2.2.2]octan-3-one, tech
(2Z)-2-[(Pyridin-3-yl)methylidene]-1-azabicyclo[2.2.2]octan-3-one, tech
Catalog # APOSOR32655-1G
Supplier:  Apollo Scientific
CAS Number:  
(2Z)-2-[(Pyridin-3-yl)methylidene]-1-azabicyclo[2.2.2]octan-3-one, tech
Catalog # APOSOR32655-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :214.263 g/mol
Storage Temperature:Ambient
CAS Number:273748-55-3
Formula:C₁₃H₁₄N₂O
MDL Number:MFCD01815113