1-[(3,4-Dichlorophenyl)methoxy]-3-[(1E)-(dimethylamino)methylidene]urea, tech
Catalog # APOSOR32610-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:290.146 g/mol
Storage Temperature:Ambient
CAS Number:338404-01-6
Formula:C₁₁H₁₃Cl₂N₃O₂
MDL Number:MFCD00139685