Order Entry
Poland
ContactUsLinkComponent
6-Hydroxy-2-phenyl-2,3-dihydropyridazin-3-one, tech
6-Hydroxy-2-phenyl-2,3-dihydropyridazin-3-one, tech
Catalog # APOSOR32572-1G
Supplier:  Apollo Scientific
CAS Number:  
6-Hydroxy-2-phenyl-2,3-dihydropyridazin-3-one, tech
Catalog # APOSOR32572-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :188.183 g/mol
Storage Temperature:Ambient
CAS Number:1698-54-0
Formula:C₁₀H₈N₂O₂
MDL Number:MFCD00010107