3-Acetyl-4-methoxy-1,2-dihydroquinolin-2-one, tech
Catalog # APOSOR32550-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:217.221 g/mol
Storage Temperature:Ambient
CAS Number:400079-79-0
Formula:C₁₂H₁₁NO₃
MDL Number:MFCD01316892