Order Entry
Poland
ContactUsLinkComponent
1,3-Dihydroimidazol-2-one 95%
Irritant
1,3-Dihydroimidazol-2-one 95%
Catalog # APOSOR323247-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1,3-Dihydroimidazol-2-one 95%
Catalog # APOSOR323247-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :84.077 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:5918-93-4
Formula:C₃H₄N₂O
MDL Number:MFCD00040252