Order Entry
Poland
ContactUsLinkComponent
(1E)-1-(Dimethylamino)-4-(1H-1,2,4-triazol-1-yl)pent-1-en-3-one, tech
(1E)-1-(Dimethylamino)-4-(1H-1,2,4-triazol-1-yl)pent-1-en-3-one, tech
Catalog # APOSOR32294-1G
Supplier:  Apollo Scientific
CAS Number:  
(1E)-1-(Dimethylamino)-4-(1H-1,2,4-triazol-1-yl)pent-1-en-3-one, tech
Catalog # APOSOR32294-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :194.234 g/mol
Storage Temperature:Ambient
CAS Number:303996-70-5
Formula:C₉H₁₄N₄O
MDL Number:MFCD00664069