Order Entry
Poland
ContactUsLinkComponent
3-Acetyl-4-hydroxy-1-(propan-2-yl)-1,2-dihydroquinolin-2-one, tech
3-Acetyl-4-hydroxy-1-(propan-2-yl)-1,2-dihydroquinolin-2-one, tech
Catalog # APOSOR32151-1G
Supplier:  Apollo Scientific
CAS Number:  
3-Acetyl-4-hydroxy-1-(propan-2-yl)-1,2-dihydroquinolin-2-one, tech
Catalog # APOSOR32151-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :245.274 g/mol
Storage Temperature:Ambient
CAS Number:860649-18-9
Formula:C₁₄H₁₅NO₃
MDL Number:MFCD05974941