Order Entry
Poland
ContactUsLinkComponent
5-((4-Methoxycyclopenta-1,3-dien-1-yl)oxy)-2-(methylthio)pyrimidine 97%
Irritant
5-((4-Methoxycyclopenta-1,3-dien-1-yl)oxy)-2-(methylthio)pyrimidine 97%
Catalog # APOSOR321312-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
5-((4-Methoxycyclopenta-1,3-dien-1-yl)oxy)-2-(methylthio)pyrimidine 97%
Catalog # APOSOR321312-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :236.29 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:1245648-98-9
Formula:C₁₁H₁₂N₂O₂S
MDL Number:MFCD12911419