Order Entry
Poland
ContactUsLinkComponent
6-(Piperidin-4-yl)-3,4-dihydroisoquinoline-2(1H)-carbaldehyde hydrochloride 97%
Irritant
6-(Piperidin-4-yl)-3,4-dihydroisoquinoline-2(1H)-carbaldehyde hydrochloride 97%
Catalog # APOSOR321240-100MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
6-(Piperidin-4-yl)-3,4-dihydroisoquinoline-2(1H)-carbaldehyde hydrochloride 97%
Catalog # APOSOR321240-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :280.793 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:1853217-54-5
Formula:C₁₅H₂₀N₂O·ClH
MDL Number:MFCD29075603