Order Entry
Poland
ContactUsLinkComponent
(S)-2-Ethylpiperidine hydrochloride 95%
Irritant
(S)-2-Ethylpiperidine hydrochloride 95%
Catalog # APOSOR321204-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
(S)-2-Ethylpiperidine hydrochloride 95%
Catalog # APOSOR321204-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :149.662 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:558479-16-6
Formula:C₇H₁₅N·ClH
MDL Number:MFCD28165866