Order Entry
Poland
ContactUsLinkComponent
3-[(4-Chlorophenyl)methyl]-4-hydroxy-1-propyl-1,2-dihydropyridin-2-one, tech
3-[(4-Chlorophenyl)methyl]-4-hydroxy-1-propyl-1,2-dihydropyridin-2-one, tech
Catalog # APOSOR32100-1G
Supplier:  Apollo Scientific
CAS Number:  
3-[(4-Chlorophenyl)methyl]-4-hydroxy-1-propyl-1,2-dihydropyridin-2-one, tech
Catalog # APOSOR32100-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :277.746 g/mol
Storage Temperature:Ambient
CAS Number:477846-40-5
Formula:C₁₅H₁₆ClNO₂
MDL Number:MFCD01568725