Order Entry
Poland
ContactUsLinkComponent
Ethyl 4-(3,3-dimethyl-2-oxoazetidin-1-yl)benzoate, tech
Ethyl 4-(3,3-dimethyl-2-oxoazetidin-1-yl)benzoate, tech
Catalog # APOSOR32079-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 4-(3,3-dimethyl-2-oxoazetidin-1-yl)benzoate, tech
Catalog # APOSOR32079-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :247.29 g/mol
Storage Temperature:Ambient
CAS Number:341965-86-4
Formula:C₁₄H₁₇NO₃
MDL Number:MFCD00793358