3-[(E)-2-(Dimethylamino)ethenyl]-5,5-dimethylcyclohex-2-en-1-one 95%
Catalog # APOSOR32021-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:193.285 g/mol
Storage Temperature:Ambient
CAS Number:128475-82-1
Formula:C₁₂H₁₉NO
MDL Number:MFCD00140800