Order Entry
Poland
ContactUsLinkComponent
2-[2-[(6-Chloro-4-pyrimidinyl)oxy]phenyl]-3-methoxy-2-propenoic acid methyl ester 97%
Irritant
2-[2-[(6-Chloro-4-pyrimidinyl)oxy]phenyl]-3-methoxy-2-propenoic acid methyl ester 97%
Catalog # APOSOR315151-100G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-[2-[(6-Chloro-4-pyrimidinyl)oxy]phenyl]-3-methoxy-2-propenoic acid methyl ester 97%
Catalog # APOSOR315151-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :320.728 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:131860-97-4
Formula:C₁₅H₁₃ClN₂O₄
MDL Number:MFCD13194671