2-(4-(Bicyclo[1.1.1]pentan-1-yl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%
Supplier: Apollo Scientific
Certificates
About this item
:
Formula:C₁₇H₂₃BO₂
MDL Number:MFCD27996124
:270.174 g/mol
Storage Temperature:Ambient
Frequently Bought Together