3-Methoxy-4-(4-methyl-1H-imidazol-1-yl)benzoic acid
Catalog # APOSOR311108-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:232.235 g/mol
Storage Temperature:Ambient
CAS Number:937026-26-1
Formula:C₁₂H₁₂N₂O₃
MDL Number:MFCD18906996