Order Entry
Poland
ContactUsLinkComponent
3-Oxo-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazine-6-carbaldehyde
Irritant
3-Oxo-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazine-6-carbaldehyde
Catalog # APOSOR310893-500MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
3-Oxo-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazine-6-carbaldehyde
Catalog # APOSOR310893-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :178.145 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:443956-11-4
Formula:C₈H₆N₂O₃
MDL Number:MFCD15526745