Order Entry
Poland
ContactUsLinkComponent
Methyl cis-2-aminocyclopentanecarboxylate hydrochloride
Methyl cis-2-aminocyclopentanecarboxylate hydrochloride
Catalog # APOSOR307667-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl cis-2-aminocyclopentanecarboxylate hydrochloride
Catalog # APOSOR307667-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :179.645 g/mol
Storage Temperature:Ambient
CAS Number:119993-55-4
Formula:C₇H₁₃NO₂·ClH
MDL Number:MFCD09997947