
4-Methoxy-2,3-dimethylpyridine
Catalog # APOSOR307220-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:137.179 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:163593-69-9
Formula:C₈H₁₁NO
MDL Number:MFCD11100693