Methyl 6-chlorospiro[3.3]heptane-2-carboxylate
Catalog # APOSOR304431-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:188.651 g/mol
Storage Temperature:Ambient
Formula:C₉H₁₃ClO₂
MDL Number:MFCD31617950