Order Entry
Poland
ContactUsLinkComponent
cis-3-Hydroxycyclobutanecarbonitrile
cis-3-Hydroxycyclobutanecarbonitrile
Catalog # APOSOR304376-1G
Supplier:  Apollo Scientific
CAS Number:  
cis-3-Hydroxycyclobutanecarbonitrile
Catalog # APOSOR304376-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :97.115 g/mol
Storage Temperature:Ambient
CAS Number:88315-79-1
Formula:C₅H₇NO
MDL Number:MFCD27952941