2-(2-Oxo-2,3,4,5-tetrahydro-1H-1-benzazepin-3-yl)-2,3-dihydro-1H-isoindole-1,3-dione
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:105260-10-4
:
Formula:C₁₈H₁₄N₂O₃
MDL Number:MFCD22689172
:306.315 g/mol
Storage Temperature:Ambient
Frequently Bought Together