4,5-Dichloro-1,2,3-dithiazol-2-ylium chloride, tech
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:75318-43-3
:
Formula:C₂Cl₂NS₂·Cl
MDL Number:MFCD03426194
:208.517 g/mol
Storage Temperature:Ambient
Frequently Bought Together