Order Entry
Poland
ContactUsLinkComponent
 
Irritant
1-Bromo-3-(2,2-dibromovinyl)-5-isopropylbenzene
Catalog # APOSOR300225-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1-Bromo-3-(2,2-dibromovinyl)-5-isopropylbenzene
Catalog # APOSOR300225-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :382.917 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:2227272-84-4
Formula:C₁₁H₁₁Br₃
MDL Number:MFCD31561256