Order Entry
Poland
ContactUsLinkComponent
2-(Bromomethyl)-1-methoxy-3-nitrobenzene
Corrosive
Irritant
2-(Bromomethyl)-1-methoxy-3-nitrobenzene
Catalog # APOSOR300181-250MG
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
Irritant
2-(Bromomethyl)-1-methoxy-3-nitrobenzene
Catalog # APOSOR300181-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :246.058 g/mol
Storage Temperature:Ambient
UN:3261
ADR:8,III
 :GHS05
 :GHS07
CAS Number:19689-86-2
Formula:C₈H₈BrNO₃
MDL Number:MFCD17168655