Order Entry
Poland
ContactUsLinkComponent
1-(4-Benzylpiperazin-1-yl)-2-(5-nitro-1H-indol-3-yl)ethane-1,2-dione
Irritant
1-(4-Benzylpiperazin-1-yl)-2-(5-nitro-1H-indol-3-yl)ethane-1,2-dione
Catalog # APOSOR28200-50MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1-(4-Benzylpiperazin-1-yl)-2-(5-nitro-1H-indol-3-yl)ethane-1,2-dione
Catalog # APOSOR28200-50MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    50 mg
  • Seq:
    2370

Specifications

About this item

 :
 :392.408 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:334973-49-8
Formula:C₂₁H₂₀N₄O₄
MDL Number:MFCD01935748